Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Biohazard Waste Containers

Biohazard Waste Containers

hvid, 34 L, 354×464 mm
Katalog # JUSU05930WH
Leverandør:   JUSTRITE MANUFACTURING
Biohazard Waste Containers
Katalog # JUSU05930WH
Leverandør:  JUSTRITE MANUFACTURING
Biohazard Waste Containers
Biohazard Waste Containers

hvid, 34 L, 354×464 mm

Price
1.580,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These leakproof sturdy steel containers with foot-operated and self-closing lid may be used to store contaminated laundry as well as other regulated waste (not for sharps). They have a biohazard bag which is held securely around the circumference of the can top.

Certifications: Comply with US OSHA Standard 29 CFR Part 1910.1030 for segregation of infectious waste. FM and TUV approved.

Specifikationer
  • Diameter×H:
    354×464 mm
  • Kapacitet:
    34 L
  • Farve:
    hvid
  • Seq:
    02
Accessories

Produktinformation

  • Katalog #
    Farve
    Kapacitet
    Tilgængelighed
    Price
  • Biohazard waste can liner bags
    Biohazard waste can liner bags
    JUSU05901
    rød
    56,8 L
    Log ind for tilgængelighed
    1 * 100 Items - 214,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.