Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
VistaRak™, 1250 µl, -, 5 racks, 192 each
Katalog # VIST4060-3004
Leverandør:   VISTALAB TECHNOLOGIES
Pipette tips
Katalog # VIST4060-3004
Leverandør:  VISTALAB TECHNOLOGIES
Pipette tips
Pipette tips
VistaRak™, 1250 µl, -, 5 racks, 192 each
Price
693,00 kr
Log ind for tilgængelighed
1 * 960 Items
Product Details & Documents
These tips to maximise the exceptional performance and benefits of ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
Specifikationer
  • Pakket:
    5 racks, 192 each
  • Steril:
    -
  • Volumen:
    1250 µl
  • Seq:
    12
  • Spidstype:
    VistaRak™