Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, Dip Slides

Pladeantal agar med TTC og MacConkey agar 3 med neutralisatorer, for totaltælling, Salmonella, Shigella og Coliform, 100
Catalog # 535317D
Supplier:   VWR Chemicals
VWR®, Dip Slides
Catalog # 535317D
Supplier:  VWR Chemicals
CAS Number:  
VWR®, Dip Slides
VWR®, Dip Slides

Pladeantal agar med TTC og MacConkey agar 3 med neutralisatorer, for totaltælling, Salmonella, Shigella og Coliform, 100

CAS Number:  
Price
1.470,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents

Dip slides for microbiological monitoring of surfaces and in liquids. Each slide has media on both sides, with a choice of the same medium, or two different media on the two sides of the slide. This allows sampling of two surfaces with every slide. Testing with dip slides is a safe, reliable, economical and fast indicator of the presence of microorganisms. Dip slides, with a push cap, have a larger contact area.

  • Leveres sterile
  • Det tager kun få sekunder at tage prøven
  • Resultaterne kan nemt aflæses efter blot 24-48 timer
Specifications

Specifications

  • Pakn:
    100
  • Beskrivelse:
    Pladeantal agar med TTC og MacConkey agar 3 med neutralisatorer, for totaltælling, Salmonella, Shigella og Coliform
  • Cat. No.:
    535317D
  • Mål:
    30 mm rør, 110 mm høj
  • Kontaktområde:
    10 cm²|10
  • Model:
    Push-on-låg
  • Bakke B×L:
    50×22 mm
  • Bladstørrelse:
    80×28 mm|80×28

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.