VWR®, Dip Slides
Pladetællingsagar med TTC og neutralisatorer, på begge sider, for totaltælling, 100
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Dip slides for microbiological monitoring of surfaces and in liquids. Each slide has media on both sides, with a choice of the same medium, or two different media on the two sides of the slide. This allows sampling of two surfaces with every slide. Testing with dip slides is a safe, reliable, economical and fast indicator of the presence of microorganisms. Dip slides, with a push cap, have a larger contact area.
- Leveres sterile
- Det tager kun få sekunder at tage prøven
- Resultaterne kan nemt aflæses efter blot 24-48 timer
Certifikater
Søg efter partinummerPack insert TDS Nutrient Agar with TTC and Malt Extract Agar Large Dip Slides (535102B) TDS Nutrient Agar with TTC and Malt Extract Agar, Pack of 10 TDS Plate Count Agar with Neutraliser Black D TDS Plate Count Agar with Neutraliser Black DI
More
Specifikationer
Specifikationer
- Pakn:100
- Beskrivelse:Pladetællingsagar med TTC og neutralisatorer, på begge sider, for totaltælling
- Cat. No.:535314D
- Mål:30 mm rør, 110 mm høj
- Kontaktområde:10 cm²|10
- Model:Push-on-låg
- Bakke B×L:50×22 mm
- Bladstørrelse:80×28 mm|80×28



