Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, Dip Slides

Næringsagar med TTC, 10
Katalog # 535306D
Leverandør:   VWR Chemicals
VWR®, Dip Slides
Katalog # 535306D
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, Dip Slides
VWR®, Dip Slides

Næringsagar med TTC, 10

CAS nummer:  
Price
148,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents

Dip slides for microbiological monitoring of surfaces and in liquids. Each slide has media on both sides, with a choice of the same medium, or two different media on the two sides of the slide. This allows sampling of two surfaces with every slide. Testing with dip slides is a safe, reliable, economical and fast indicator of the presence of microorganisms. Dip slides, with a push cap, have a larger contact area.

  • Leveres sterile
  • Det tager kun få sekunder at tage prøven
  • Resultaterne kan nemt aflæses efter blot 24-48 timer
Specifikationer

Specifikationer

  • Pakn:
    10
  • Beskrivelse:
    Næringsagar med TTC
  • Cat. No.:
    535306D
  • Mål:
    24 mm rør, 92 mm høj
  • Kontaktområde:
    10 cm²|10
  • Model:
    Skruelåg
  • Bakke B×L:
    50×15 mm
  • Bladstørrelse:
    72×20 mm|80×28

Specifikationer