VWR®, Dip Slides
Pladeantal agar og VRBGA med neutralisatorer til Staphylococcus aureus og Enterobacteriaceae, 10
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
Dip slides for microbiological monitoring of surfaces and in liquids. Each slide has media on both sides, with a choice of the same medium, or two different media on the two sides of the slide. This allows sampling of two surfaces with every slide. Testing with dip slides is a safe, reliable, economical and fast indicator of the presence of microorganisms. Dip slides, with a push cap, have a larger contact area.
- Leveres sterile
- Det tager kun få sekunder at tage prøven
- Resultaterne kan nemt aflæses efter blot 24-48 timer
Documents
Certifikater
Søg efter partinummerPack insert TDS Nutrient Agar with TTC and Malt Extract Agar Large Dip Slides (535102B) TDS Nutrient Agar with TTC and Malt Extract Agar, Pack of 10 TDS Plate Count Agar with Neutraliser Black D TDS Plate Count Agar with Neutraliser Black DI
Datasheet
More
SDS_DA_DK_VWRC53510.pdf SDS_DA_DK_VWRC53511.pdf SDS_DA_DK_VWRC53512.pdf SDS_DA_DK_VWRC535132H.pdf SDS_DA_DK_VWRC535134J.pdf
SDS
More
Spec VWRC535314D.pdf spec_VWRC535124H spec_VWRC535134J spec_VWRC535142J spec_VWRC535202E
Product specification
Specifikationer
Specifikationer
- Pakn:10
- Beskrivelse:Pladeantal agar og VRBGA med neutralisatorer til Staphylococcus aureus og Enterobacteriaceae
- Cat. No.:535172P
- Mål:30 mm rør, 110 mm høj
- Kontaktområde:10 cm²|10
- Model:Push-on-låg
- Bakke B×L:50×22 mm
- Bladstørrelse:80×28 mm|80×28



