Orders LinkContactUsLinkComponent
Latex tests, Dryspot
Dryspot Legionella Pneumophila Serogroup, 60
  OXOIDR0210M
Latex tests, Dryspot
  OXOIDR0210M
 :  
Latex tests, Dryspot
Dryspot Legionella Pneumophila Serogroup, 60
 :  
2.700,00 kr
1 * 60 Items
Dry latex test designed to demonstrate by slide agglutination strains possessing a serogroup antigen.
  • Simplified room temperature storage allows storage for up to two years
  • Blue latex particles are sensitized with reactive (positive) and non-reactive (negative) antibodies and dried onto reaction cards
  • Pakn:
    60
  • Beskrivelse:
    Dryspot Legionella Pneumophila Serogroup