Latex tests, Dryspot
Dryspot Legionella Pneumophila Serogroup, 60
Dry latex test designed to demonstrate by slide agglutination strains possessing a serogroup antigen.
- Simplified room temperature storage allows storage for up to two years
- Blue latex particles are sensitized with reactive (positive) and non-reactive (negative) antibodies and dried onto reaction cards
- Pakn:60
- Beskrivelse:Dryspot Legionella Pneumophila Serogroup