Sediment testers for milk
500 ml, SEDILAB-E
Katalog # 614-0196
Leverandør: GERBER FUNKE
Gå til:
Product Details & Documents
SEDILAB: Manual sediment tester. With clamp for attaching to tables.
SEDILAB-E: Pump design sediment tester for serial testing of liquids for particle contamination, particularly for sediment testing of milk. Splash-proof design. Approx. 800 samples per hour. Sharply defined sediment images.
Pump design sediment tester for serial testing of liquids for particle contamination, particularly for sediment testing of milk.
Specifikationer
- Model:SEDILAB-E
- Kapacitet:500 ml