Orders LinkContactUsLinkComponent
Sediment testers for milk
500 ml, SEDILAB-E
Katalog # 614-0196
Leverandør:   GERBER FUNKE
Sediment testers for milk
Katalog # 614-0196
Leverandør:  GERBER FUNKE
Sediment testers for milk
Sediment testers for milk
500 ml, SEDILAB-E
Price
14.330,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
SEDILAB: Manual sediment tester. With clamp for attaching to tables.
SEDILAB-E: Pump design sediment tester for serial testing of liquids for particle contamination, particularly for sediment testing of milk. Splash-proof design. Approx. 800 samples per hour. Sharply defined sediment images.
Pump design sediment tester for serial testing of liquids for particle contamination, particularly for sediment testing of milk.
Specifikationer
  • Model:
    SEDILAB-E
  • Kapacitet:
    500 ml