Pipette tips, Matrix™ SerialMate™
Filtered, 5,0 - 220 µl, +, +, 45,72 mm, Matrix™ SerialMate™, 20 racks, 96 each
Katalog # MATR5545
Leverandør: THERMO MATRIX TECHNOLOGIES
Pipette tips, Matrix™ SerialMate™
Katalog # MATR5545
Leverandør: THERMO MATRIX TECHNOLOGIES
Gå til:
Product Details & Documents
Pipette tips for use with the Matrix™ SerialMate™ pipetting station. Compatible with 96-well plates.
Specifikationer
- Pakket:20 racks, 96 each
- Steril:+
- Volumen:5,0 - 220 µl
- Længde:45,72 mm
- Spidstype:Filtered
- Til brug med:Matrix™ SerialMate™
- Filter:+
- Cat. No.:MATR5545
- Miljøcertificering:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Lav fremstillingspåvirkning