Overflow volumetric flasks, class B
97 ml
Catalog # HECH42115024
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
DURAN® borosilicate glass 3.3.
- Acid-proof and alkali-resistant markings
- Very good temperature resistance
- Very good chemical resistance
For water analysis.
Specifications
- Kapacitet:97 ml
- Seq:04