Overflow volumetric flasks, class B
250 ml
Katalog # HECH42115048
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
DURAN® borosilicate glass 3.3.
- Acid-proof and alkali-resistant markings
- Very good temperature resistance
- Very good chemical resistance
For water analysis.
Specifikationer
- Kapacitet:250 ml
- Seq:07