Cyclohexan ≥99.8%, Laboratory Reagent
Metal can, 10 L
Leverandør: Honeywell Chemicals
Cyclohexan ≥99.8%, Laboratory Reagent
Katalog # C100307-10L
Leverandør: Honeywell Chemicals
CAS nummer: 110-82-7
Gå til:
Product Details & Documents
- ≥95% for general laboratory use.
Beilstein Number: 1900225
RTECS Number: GU6300000
UNSPSC Code: 12191504
Synonymer: Hexahydrobenzene, Hexamethylene
- CAS nummer:110-82-7
- Formel:C₆H₁₂
- MDL nummer:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Kogende Pt:8,3 %(V)
- Smeltende Pt:6,4 °C
- Tæthed:0,78 g/cm³ (20 °C)
- Flash Pt:–18 °C
- Flash Pt:-18 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-806-2
- FN:1145
- ADR:3,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance (form):complying
- Appearance (color):complying
- Purity (GC):Min. 99.80 Fl.%
- Non-volatile matter:Max. 0.005 %
- IR spectrum:complying
- Vapor Density:2.9 (vs air)
- Vapor Pressure:77 mmHg ( 20 °C)
- Autoignition Temperature:500 °F
- Refractive Index:n20/D 1.426 (lit.)
Specifikationer
- Pakning:Metal can
- Pakn:10 L