Kieselgel, Supelco®
5 kg
12479-5KG
: Merck
- :63231-67-4
- :SiO₂
- :MFCD00011232
- :60.08 g/mol
- :2230 °C (1013 hPa)
- :1710 °C
- :M-Clarity™ quality level = MQ100
- :Stue temperatur
- :01-2119379499-16
- Pakn:5 kg