Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Threaded fittings

Threaded fittings

Tubing nozzles, fixed, external thread, nozzle-Ø 12 mm, NW 7,5 mm, PP, 9,5 mm
Katalog # BURK8545-3812
Leverandør:   BURKLE
Threaded fittings
Katalog # BURK8545-3812
Leverandør:  BURKLE
Threaded fittings
Threaded fittings

Tubing nozzles, fixed, external thread, nozzle-Ø 12 mm, NW 7,5 mm, PP, 9,5 mm

Price
14,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Gevind:
    9,5 mm
  • Beskrivelse:
    Tubing nozzles, fixed, external thread, nozzle-Ø 12 mm, NW 7,5 mm
  • Materiale:
    PP