Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for panthera

Accessories for panthera

Reticle with crossed double scale 100 divisions in 10 mm (Ø 25 mm)
Katalog # MOTI1101001401861
Leverandør:   MOTIC
Accessories for panthera
Katalog # MOTI1101001401861
Leverandør:  MOTIC
Accessories for panthera
Accessories for panthera

Reticle with crossed double scale 100 divisions in 10 mm (Ø 25 mm)

Price
379,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Reticle with crossed double scale 100 divisions in 10 mm (Ø 25 mm)
  • Seq:
    21