Phenol rød natrium salt
Glass bottle for solids, 1 kg
Leverandør: Thermo Fisher Scientific
Phenol rød natrium salt
Katalog # B21948.A1
Leverandør: Thermo Fisher Scientific
CAS nummer: 34487-61-1
Gå til:
Product Details & Documents
CAS No.: 34487-61-1 Molecular Formula: C19H13NaO5S Formula Weight: 376.37 Melting Point: 285° dec. Beilstein Reference: 6260026 Merck Reference: 14,7247 MDL No.: MFCD00066901
Synonymer: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide
- CAS nummer:34487-61-1
- Formel:C₁₉H₁₃NaO₅S
- MDL nummer:MFCD00066901
- Molecular Weight:376.35 g/mol
- Merck Index:14,7247
- Smeltende Pt:285 °C (dec.)
- Opbevaringstemperatur:Stue temperatur
- EINECS:252-057-8
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg



