Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Natriumhydroxid, pellets NF, Multi-Compendial, J.T.Baker®

PE drum, open head, 80 lb
Catalog # 3718-20
Corrosive
Supplier:   Avantor
Natriumhydroxid, pellets NF, Multi-Compendial, J.T.Baker®
Catalog # 3718-20
Supplier:  Avantor
CAS Number:  1310-73-2
Natriumhydroxid, pellets NF, Multi-Compendial, J.T.Baker®
Natriumhydroxid, pellets NF, Multi-Compendial, J.T.Baker®

PE drum, open head, 80 lb

CAS Number:  1310-73-2
Price
12.850,00 kr
Log in for availability
1 * 80 lb
Product Details & Documents

Meets N.F. Requirements, Meets J.P. Chemical Specifications, Meets E.P. Chemical Specifications, Meets B.P. Chemical Specifications, GMP Manufactured Product

  • GMP Manufactured Product
  • Bulk Pharmaceutical Chemical

 : For Manufacturing, processing or repackaging

Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:323 °C
  • Density:2,13 g/cm³ (20 °C)
  • EINECS:215-185-5
  • ADR:8,II
  •  :Management of Change (MOC) category = R
  • Storage Temperature:Stue temperatur
  • UN:1823
  • REACH:01-2119457892-27
Specifications

Specification Test Results

  • NF - Identification A (Sodium):
    Passes Test
  • NF - Identification B (pH):
    ≥ 11.0
  • NF - Insoluble Substances and Organic Matter:
    Passes Test
  • NF - Potassium (K):
    ≤ 0.5 %
  • NF - Carbonate (as Na₂CO₃):
    ≤ 3.0 %
  • NF - Total Alkali:
    95.0 - 100.5 %
  • NF - Content of Sodium:
    54.0 - 59.8 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Carbonate (as Na₂CO₃):
    ≤ 2.0 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤ 200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤ 200 ppm
  • EP/BP - Iron (Fe):
    ≤ 10 ppm
  • EP/BP - Assay (NaOH):
    97.0 - 100.5 %
  • JP - Assay:
    95.0 - 101.0 %
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤ 0.050 %
  • JP - Potassium (K):
    Passes Test
  • JP - Carbonate (as Na₂CO₃):
    ≤ 2.0 %
  • No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
    ≤2.0%
  • Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.:
    Passes Test

Specifications

  • Pakning:
    PE drum, open head
  • Pakn:
    80 lb

Specifications