2-Propanol USP, J.T.Baker®
20 L
Leverandør: Avantor
2-Propanol USP, J.T.Baker®
Katalog # 9080-07
Leverandør: Avantor
CAS nummer: 67-63-0
Gå til:
Product Details & Documents
Synonymer: Isopropylalkohol
- CAS nummer:67-63-0
- Formel:(CH₃)₂CHOH
- MDL nummer:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Kogende Pt:82 °C (1013 hPa)
- Smeltende Pt:–89 °C
- Tæthed:0,786 g/cm³ (20 °C)
- Flash Pt:12 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-661-7
- FN:1219
- ADR:3,II
- NÅ:01-2119457558-25
Documents
Certifikater
Søg efter partinummerP20_BAKR9080-03_SPEC_EN.pdf spec_BAKR9080-01 spec_BAKR9080-R
Produktspecifikation
Specifikationer
Specifikationstestresultater
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Specific Gravity at 25°/25°C:0.783 - 0.787
- USP - Refractive Index, n²ºD:1.376 - 1.378
- USP - Acidity:Passes Test
- USP - Nonvolatile Residue:≤0.005 %
- USP - Assay:≥99.0 %
- USP - Water (H₂O):≤0.5 %
- USP - Volatile Impurities - Ethyl ether:≤0.1 %
- USP - Volatile Impurities - Acetone:≤0.1 %
- USP - Volatile Impurities - Diisopropyl ether:≤0.1 %
- USP - Volatile Impurities - 1-Propanol:≤0.1 %
- USP - Volatile Impurities - 2-Butanol:≤0.1 %
- USP - Volatile Impurities - Total:≤1.0 %
- USP - Benzene:≤2 ppm
Specifikationer
- Pakn:20 L
- Cat. No.:9080-07
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø



