Orders LinkContactUsLinkComponent
Insoles, grey
Insoles, grey
37, grå
Katalog # ABEB3551/37
Leverandør:   Abeba
Insoles, grey
Katalog # ABEB3551/37
Leverandør:  Abeba
Insoles, grey
Insoles, grey
37, grå
Price
105,00 kr
Log ind for tilgængelighed
1 * 1 Par
Product Details & Documents
Replaceable insole for shoes with air cushion sole.
  • Anti-static
Colour: Grey
Certifications: EN 61340 (ESD)
Documents
Specifikationer
  • Skostørrelse:
    37
  • Farve:
    grå
  • Seq:
    03