Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Threaded fittings

Threaded fittings

Screw joint with inner thread NW 21,5 mm, PP, 19,05 mm
Katalog # BURK8510-0034
Leverandør:   BURKLE
Threaded fittings
Katalog # BURK8510-0034
Leverandør:  BURKLE
Threaded fittings
Threaded fittings

Screw joint with inner thread NW 21,5 mm, PP, 19,05 mm

Price
190,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Gevind:
    19,05 mm
  • Beskrivelse:
    Screw joint with inner thread NW 21,5 mm
  • Materiale:
    PP
  • Seq:
    46