Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
500 ml, 22 mm, 60×70×160 mm
SEMA2025
: SEMADENI
HDPE, translucent, with thread.
- Good chemical resistance
- Good temperature resistance
: Screw caps must be ordered separately.
- Kapacitet:500 ml
- B×D×H:60×70×160 mm
- Hals indv. Ø:22 mm