Orders LinkContactUsLinkComponent
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
24/29 NS, 24/29 NS, 150 mm
Katalog # 201-0598
Leverandør:   SciLabware
Adapters, receiver, vacuum, Quickfit®
Katalog # 201-0598
Leverandør:  SciLabware
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
24/29 NS, 24/29 NS, 150 mm
Price
810,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass, bent.
  • Easy connection to flexible tubing via detachable plastic screw thread connector
Adapter with a side arm for connection to a vacuum supply.
Specifikationer
  • Konusstr.:
    24/29 NS
  • Kappestr.:
    24/29 NS
  • Samlet længde:
    150 mm