Orders LinkContactUsLinkComponent
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
14/23 NS, 14/23 NS, 110 mm
Katalog # 201-0597
Leverandør:   SciLabware
Adapters, receiver, vacuum, Quickfit®
Katalog # 201-0597
Leverandør:  SciLabware
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
14/23 NS, 14/23 NS, 110 mm
Price
741,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass, bent.
  • Easy connection to flexible tubing via detachable plastic screw thread connector
Adapter with a side arm for connection to a vacuum supply.
Specifikationer
  • Konusstr.:
    14/23 NS
  • Kappestr.:
    14/23 NS
  • Samlet længde:
    110 mm