Adapters, delivery, with vent, QUICKFIT®
14/23 NS, 14/23 NS
Katalog # 271-0008
Leverandør: SciLabware
Gå til:
Product Details & Documents
PYREX®, borosilicate glass, bend.
- Avoids the problems of 'closed systems'
With a side arm for venting purposes, not for connection to a vacuum source. For adapters with vacuum connection, see 201-8653.
Specifikationer
- Konusstr.:14/23 NS
- Kappestr.:14/23 NS
- Seq:01