Orders LinkContactUsLinkComponent
Adapters, delivery, with vent, QUICKFIT®
14/23 NS, 14/23 NS
Katalog # 271-0008
Leverandør:   SciLabware
Adapters, delivery, with vent, QUICKFIT®
Katalog # 271-0008
Leverandør:  SciLabware
Adapters, delivery, with vent, QUICKFIT®
Adapters, delivery, with vent, QUICKFIT®
14/23 NS, 14/23 NS
Price
456,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass, bend.
  • Avoids the problems of 'closed systems'
With a side arm for venting purposes, not for connection to a vacuum source. For adapters with vacuum connection, see 201-8653.
Specifikationer
  • Konusstr.:
    14/23 NS
  • Kappestr.:
    14/23 NS
  • Seq:
    01