Spherical ground joints, Rotulex®
O-ring PTFE, for joint size 35, Sfæriske samlinger RM:RF35
Katalog # BIBBRO35
Leverandør: SciLabware
Gå til:
Product Details & Documents
Made of borosilicate glass.
- All glass and PTFE pathway is highly resistant to chemical attack
- Can be used at temperatures up to 200 °C
- System comprises male ball joint with PTFE covered 'O' ring which fits into a female cup joint
- Joint can be retained together with spring loaded metal clip
Rotulex® offers a grease free spherical joint.
Ordering information: Component parts should be ordered separately .
Specifikationer
- Til:Sfæriske samlinger RM:RF35
- Model:for joint size 35
- Beskrivelse:O-ring PTFE
- Seq:19