Orders LinkContactUsLinkComponent
Spherical ground joints, Rotulex®
Spherical ground joints, Rotulex®
O-ring PTFE, for joint size 35, Sfæriske samlinger RM:RF35
Katalog # BIBBRO35
Leverandør:   SciLabware
Spherical ground joints, Rotulex®
Katalog # BIBBRO35
Leverandør:  SciLabware
Spherical ground joints, Rotulex®
Spherical ground joints, Rotulex®
O-ring PTFE, for joint size 35, Sfæriske samlinger RM:RF35
Price
276,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Made of borosilicate glass.
  • All glass and PTFE pathway is highly resistant to chemical attack
  • Can be used at temperatures up to 200 °C
  • System comprises male ball joint with PTFE covered 'O' ring which fits into a female cup joint
  • Joint can be retained together with spring loaded metal clip
Rotulex® offers a grease free spherical joint.
Ordering information: Component parts should be ordered separately .
Specifikationer
  • Til:
    Sfæriske samlinger RM:RF35
  • Model:
    for joint size 35
  • Beskrivelse:
    O-ring PTFE
  • Seq:
    19