Orders LinkContactUsLinkComponent
Tweezers
Tweezers
Straight, pointed tips, 95 mm
Katalog # KOEH3-1232
Tweezers
Katalog # KOEH3-1232
Tweezers
Straight, pointed tips, 95 mm
Price
247,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Lightweight aluminium tweezers for universal use.
  • Fully seamless
  • Anti-magnetic
  • Not corrosive
  • Packed in protective tube
Specifikationer
  • Model:
    Straight, pointed tips
  • Længde:
    95 mm