Tweezers
Straight, round (blunt) tips, 95 mm
Katalog # KOEH3-1230
Leverandør: KOEHLER TECHNISCHE PRODUKTEN
Gå til:
Product Details & Documents
Lightweight aluminium tweezers for universal use.
- Fully seamless
- Anti-magnetic
- Not corrosive
- Packed in protective tube
Specifikationer
- Model:Straight, round (blunt) tips
- Længde:95 mm