Orders LinkContactUsLinkComponent
Accessories for wire baskets
Accessories for wire baskets
275×375 mm, mesh dimensions: 20×20×3 mm
Katalog # BOCH17321
Leverandør:   BOCHEM
Accessories for wire baskets
Katalog # BOCH17321
Leverandør:  BOCHEM
Accessories for wire baskets
Accessories for wire baskets
275×375 mm, mesh dimensions: 20×20×3 mm
Price
2.900,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    275×375 mm, mesh dimensions: 20×20×3 mm
  • Seq:
    11