Orders LinkContactUsLinkComponent
Accessories for wire baskets
Accessories for wire baskets
575×375 mm, mesh dimensions: 10×10×3 mm
Katalog # BOCH17315
Leverandør:   BOCHEM
Accessories for wire baskets
Katalog # BOCH17315
Leverandør:  BOCHEM
Accessories for wire baskets
Accessories for wire baskets
575×375 mm, mesh dimensions: 10×10×3 mm
Price
3.230,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    575×375 mm, mesh dimensions: 10×10×3 mm
  • Seq:
    13