Skalpeller
1, 150 mm, Skalpel, med blad monteret, blå
Katalog # BOCH12640
Leverandør: BOCHEM
Gå til:
Product Details & Documents
These mounted scalpels feature a steel blade and a coloured handle made of impact resistant PS.
- Not sterile
- Blades are exchangeable
- Spare blades available (set of 10 pieces)
Specifikationer
- Model:1
- Beskrivelse:Skalpel, med blad monteret, blå
- Længde:150 mm