Orders LinkContactUsLinkComponent
Pharmacy scoops
Pharmacy scoops
100 ml, 220 mm, 50 mm
Catalog # BOCH8830
Supplier:   BOCHEM
Pharmacy scoops
Catalog # BOCH8830
Supplier:  BOCHEM
Pharmacy scoops
Pharmacy scoops
100 ml, 220 mm, 50 mm
Price
957,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel 18/10.
  • Soldered handle
Specifications
  • Ø:
    50 mm
  • Kapacitet:
    100 ml
  • Længde:
    220 mm
  • Seq:
    01