Support clips
Support clip with fitting, O. Ø 100 - 104 mm
Katalog # BOCH597
Leverandør: BOCHEM
Gå til:
Product Details & Documents
These support clips are coated with ceramic inside.
- For tubes and cylindrical vessels
- With fitting
Bestillingsoplysninger: The shafts (rods) need to be ordered separately.
Specifikationer
- Beskrivelse:Support clip with fitting, O. Ø 100 - 104 mm