Support clips
Support clip with fitting, O. Ø 31 - 35 mm
Katalog # BOCH591
Leverandør: BOCHEM
Gå til:
Product Details & Documents
These support clips are coated with ceramic inside.
- For tubes and cylindrical vessels
- With fitting
Ordering information: The shafts (rods) need to be ordered separately.
Specifikationer
- Beskrivelse:Support clip with fitting, O. Ø 31 - 35 mm