Orders LinkContactUsLinkComponent
Racks for freezer
Racks for freezer
13 (1×13), 32 mm, 140×140×479 mm
Katalog # RATI5400034
Leverandør:   RATIOLAB
Racks for freezer
Katalog # RATI5400034
Leverandør:  RATIOLAB
Racks for freezer
13 (1×13), 32 mm, 140×140×479 mm
Price
kr1,200.00
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
For the storing of cryoboxes in freezer cabinets and chests.
  • Robust construction
  • Handles for convenient transport
  • Made of stainless steel with surfaces for perfect cleaning
Leveringsinformation: Cryo racks are supplied with shelf sizes for any standard cryoboxes.
Specifikationer
  • Rum:
    13 (1×13)
  • B×D×H udv.:
    140×140×479 mm
  • Seq:
    12
  • Cryobox højde:
    32 mm