Test tubes, disposable, without rim
98/100 mm, 16 mm, 0,5 mm
HECH42769045
Glas, clear.
- Excellent chemical resistance
- Good temperature resistance
- Udv.Ø:16 mm
- Vægtykkelse:0,5 mm
- Længde:98/100 mm