Sample cups for analysers, ratiolab®
For Gemsaec G1 analysers, 0,5 ml
Katalog # RATI5700200
Leverandør: RATIOLAB
Gå til:
Product Details & Documents
Made of clear PS.
- Good chemical resistance
- Good temperature resistance
Analysis beakers offer the highest precision for use with robotic analytical systems.
Specifikationer
- Kapacitet:0,5 ml
- Beskrivelse:For Gemsaec G1 analysers
- Seq:01