Orders LinkContactUsLinkComponent
Racks for freezer
Racks for freezer
12 (4×3), 100 mm, 566×140×314 mm
Katalog # RATI5400040
Leverandør:   RATIOLAB
Racks for freezer
Katalog # RATI5400040
Leverandør:  RATIOLAB
Racks for freezer
12 (4×3), 100 mm, 566×140×314 mm
Price
1.350,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
For the storing of cryoboxes in freezer cabinets and chests.
  • Robust construction
  • Handles for convenient transport
  • Made of stainless steel with surfaces for perfect cleaning
Leveringsinformation: Cryo racks are supplied with shelf sizes for any standard cryoboxes.
Specifikationer
  • Rum:
    12 (4×3)
  • B×D×H udv.:
    566×140×314 mm
  • Seq:
    03
  • Cryobox højde:
    100 mm