Poser, biologisk risikomateriale
Disposable waste bags, with indicator field, PP, 72 L, 350 stk./pakke, 1100×700 mm
Katalog # RATI7002300
Leverandør: RATIOLAB
Gå til:
Product Details & Documents
These disposable waste bags are made from strong PP film with a broad seam at the bottom. Intended for the sterilisation and disposal of hazardous and contaminated labware.
- Rectangle white indicator patch field turns into dark when autoclaved
- Bags must not be completely closed while autoclaving
Available with biohazard sticker and safety imprint in English, German, French and Spanish language.
Specifikationer
- Pakket:350 stk./pakke
- Kapacitet:72 L
- Beskrivelse:Disposable waste bags, with indicator field
- L×B:1100×700 mm
- Materiale:PP
- Seq:04
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



