Poser, biologisk risikomateriale
Disposable waste bags, with indicator field, PP, 6 L, 500 stk./pakke, 500×300 mm
Katalog # RATI7002100
Leverandør: RATIOLAB
Gå til:
Product Details & Documents
These disposable waste bags are made from strong PP film with a broad seam at the bottom. Intended for the sterilisation and disposal of hazardous and contaminated labware.
- Rectangle white indicator patch field turns into dark when autoclaved
- Bags must not be completely closed while autoclaving
Available with biohazard sticker and safety imprint in English, German, French and Spanish language.
Specifikationer
- Pakket:500 stk./pakke
- Kapacitet:6 L
- Beskrivelse:Disposable waste bags, with indicator field
- L×B:500×300 mm
- Materiale:PP
- Seq:01



