Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for centrifuges

Accessories for centrifuges

Adapter for 12×2,7 - 6 ml tubes (11/12×100 mm)
Katalog # HETT5229
   Lav fremstillingspåvirkning
Leverandør:   Hettich
Accessories for centrifuges
Katalog # HETT5229
Leverandør:  Hettich
   Lav fremstillingspåvirkning
Accessories for centrifuges
Accessories for centrifuges

Adapter for 12×2,7 - 6 ml tubes (11/12×100 mm)

Price
653,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Adapter for 12×2,7 - 6 ml tubes (11/12×100 mm)
  • Seq:
    26
  • Cat. No.:
    HETT5229
  • Miljøcertificering:
    ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • Environmentally Preferable:
    Lav fremstillingspåvirkning
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.