Orders LinkContactUsLinkComponent
Accessories for centrifuges
Accessories for centrifuges
Adapter for 12×1,5/2 ml tubes (11×38 mm)
Katalog # HETT5273
   Lav fremstillingspåvirkning
Leverandør:   Hettich
Accessories for centrifuges
Katalog # HETT5273
Leverandør:  Hettich
   Lav fremstillingspåvirkning
Accessories for centrifuges
Accessories for centrifuges
Adapter for 12×1,5/2 ml tubes (11×38 mm)
Price
912,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Adapter for 12×1,5/2 ml tubes (11×38 mm)
  • Seq:
    24
  • Cat. No.:
    HETT5273
  • Miljøcertificering:
    ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • Environmentally Preferable:
    Lav fremstillingspåvirkning