Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for centrifuges

Accessories for centrifuges

Adapter til 7×50 ml Falconrør m. konisk bund, 30×96,5 mm
Catalog # 521-1267
   Lav fremstillingspåvirkning
Supplier:   Hettich
Accessories for centrifuges
Catalog # 521-1267
Supplier:  Hettich
   Lav fremstillingspåvirkning
Accessories for centrifuges
Accessories for centrifuges

Adapter til 7×50 ml Falconrør m. konisk bund, 30×96,5 mm

Price
1.470,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Adapter til 7×50 ml Falconrør m. konisk bund, 30×96,5 mm
  • Seq:
    40
  • Cat. No.:
    521-1267
  • Miljøcertificering:
    ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • Environmentally Preferable:
    Lav fremstillingspåvirkning
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.