Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Accessories for centrifuges

Adapter for 5×4 - 8,2 ml tubes (15×92 mm)
  HETT5279
   Lav fremstillingspåvirkning
 :   Hettich
Accessories for centrifuges
  HETT5279
 :  Hettich
   Lav fremstillingspåvirkning
Accessories for centrifuges
Accessories for centrifuges

Adapter for 5×4 - 8,2 ml tubes (15×92 mm)

 
856,00 kr
   
1 * 1 Items
 
 
  • Beskrivelse:
    Adapter for 5×4 - 8,2 ml tubes (15×92 mm)
  • Seq:
    36
  • Cat. No.:
    HETT5279
  • Miljøcertificering:
    ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • Environmentally Preferable:
    Lav fremstillingspåvirkning