Jump to:
Product Details & Documents
Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
- CAS Number:7772-98-7
- Formula:Na₂S₂O₃
- MDL Number:MFCD00003499
- Molecular Weight:158.11 g/mol
- Merck Index:12,08844
- Boiling Pt:100 °C (1013 hPa)
- Melting Pt:48 °C
- Density:1,66 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Stue temperatur
- EINECS:231-867-5
Documents
Certificates
Search by Lot NumberMERC1.06512.2500_TDS_EN ProductPage_106512
Product specification
msds_106512
SDS
Specifications
Specifications
- Pakning:Fibre carton
- Pakn:25 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



