Chloroform ≥99.5% stabilised
Glass bottle, 500 mL
Leverandør: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Katalog # C2432-500ML
Leverandør: Honeywell Chemicals
CAS nummer: 67-66-3
Gå til:
Product Details & Documents
Beilstein Number: 1731042
RTECS Number: FS9100000
UNSPSC Code: 12191502
Synonymer: Trichlormethan
- CAS nummer:67-66-3
- Formel:CHCl₃
- MDL nummer:MFCD00000826
- Molecular Weight:119.38 g/mol
- Kogende Pt:61 °C (1,013 hPa)
- Smeltende Pt:–61,3 °C
- Smeltende Pt:-61,3 °C
- Tæthed:1,485 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-663-8
- FN:1888
- ADR:6.1,III
- :stabiliseret med amylen 100-200 ppm
Documents
Specifikationer
Specifikationstestresultater
- Vapor Pressure:160 mmHg ( 20 °C)
- NMR-spectrum:complying
- Assay (GC):Min. 99.50 %
- 2-methyl-2-buten (GC):20 - 200 ppm
- Refractive Index:n20/D 1.445 (lit.)
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL



