Chloroform ≥99.5% stabilised
Glass bottle, 25 mL
Beilstein Number: 1731042
RTECS Number: FS9100000
UNSPSC Code: 12191502
: Trichlormethan
- :67-66-3
- :CHCl₃
- :MFCD00000826
- :119.38 g/mol
- :61 °C (1,013 hPa)
- :–61,3 °C
- :-61,3 °C
- :1,485 g/cm³ (20 °C)
- :Stue temperatur
- :200-663-8
- :1888
- :6.1,III
- :stabiliseret med amylen 100-200 ppm
- Vapor Pressure:160 mmHg ( 20 °C)
- NMR-spectrum:complying
- Assay (GC):Min. 99.50 %
- 2-methyl-2-buten (GC):20 - 200 ppm
- Refractive Index:n20/D 1.445 (lit.)
- Pakning:Glass bottle
- Pakn:25 mL



