Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumsilikat solution, Reagent Grade

Natriumsilikat solution, Reagent Grade

Plastic bottle, 3 L
Katalog # 338443-3L
Irriterende
Ætsende
Leverandør:   Honeywell Chemicals
Natriumsilikat solution, Reagent Grade
Katalog # 338443-3L
Leverandør:  Honeywell Chemicals
CAS nummer:  1344-09-8
Natriumsilikat solution, Reagent Grade

Plastic bottle, 3 L

CAS nummer:  1344-09-8
Price
1.360,00 kr
Log ind for tilgængelighed
1 * 3 L
Product Details & Documents
  • ≥95% for general laboratory use.
Synonymer:  Vandglas
  • CAS nummer:1344-09-8
  • Formel:Na₂O(SiO₂)x·xH₂O
  • MDL nummer:MFCD00003492
  • Kogende Pt:100 °C
  • Opbevaringstemperatur:Stue temperatur
  • ADR:8,III
Specifikationer

Specifikationstestresultater

  • Assay of NaOH:
    13.4 - 14.4 %
  • Assay of SiO₂:
    25.7 - 27.8 %
  • Composition:
    SiO₂, ~26.5%

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    3 L

Specifikationer