Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumsilikat solution, Reagent Grade

Natriumsilikat solution, Reagent Grade

Plastic bottle, 1 L
Katalog # 338443-1L
Irriterende
Ætsende
Leverandør:   Honeywell Chemicals
Natriumsilikat solution, Reagent Grade
Katalog # 338443-1L
Leverandør:  Honeywell Chemicals
CAS nummer:  1344-09-8
Natriumsilikat solution, Reagent Grade

Plastic bottle, 1 L

CAS nummer:  1344-09-8
Price
477,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
  • ≥95% for general laboratory use.
Synonymer:  Vandglas
  • CAS nummer:1344-09-8
  • Formel:Na₂O(SiO₂)x·xH₂O
  • MDL nummer:MFCD00003492
  • Kogende Pt:100 °C
  • Opbevaringstemperatur:Stue temperatur
  • ADR:8,III
Specifikationer

Specifikationstestresultater

  • Assay of NaOH:
    13.4 - 14.4 %
  • Assay of SiO₂:
    25.7 - 27.8 %
  • Composition:
    SiO₂, ~26.5%

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    1 L

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.