Anisol 99%, Reagent Grade
Glass bottle, 2,5 L
Leverandør: Honeywell Chemicals
Anisol 99%, Reagent Grade
Katalog # 123226-2.5L
Leverandør: Honeywell Chemicals
CAS nummer: 100-66-3
Gå til:
Product Details & Documents
- ≥95% for general laboratory use.
Synonymer: Methyl phenyl ether, Methoxybenzene
- CAS nummer:100-66-3
- Formel:C₇H₈O
- MDL nummer:MFCD00008354
- Molecular Weight:108.14 g/mol
- Merck Index:13,00672
- Kogende Pt:154 °C (1,013 hPa)
- Smeltende Pt:–37 °C
- Smeltende Pt:-37 °C
- Tæthed:1,00 g/cm³ (20 °C)
- Flash Pt:43 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-876-1
- FN:2222
- ADR:3,III
Documents
sds_123226
SDS
Specifikationer
Specifikationstestresultater
- Vapor Density:3.7 (vs air)
- Assay (GC):Min. 98.5 %
- Identity (IR):complying
- Vapor Pressure:10 mmHg ( 42.2 °C)
- Autoignition Temperature:887 °F
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L
- Cat. No.:123226-2.5L
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



