Tetrachlorethylen ≥99.5% stabilised
Glass bottle, 6 L
Beilstein Number: 1361721
RTECS Number: KX3850000
UNSPSC Code: 12190000
: Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
- :127-18-4
- :C₂Cl₄
- :MFCD00000834
- :165.83 g/mol
- :12,09332
- :120…122 °C (1,013 hPa)
- :120-122 °C (1,013 hPa)
- :–22 °C
- :-22 °C
- :1,622…1.624 g/cm3 (20 °C)
- :Stue temperatur
- :204-825-9
- :1897
- :6.1,III
Documents
sds_16211
SDS
- Vapor Density:5.83 (vs air)
- Contains:stabilizer
- Assay:Min. 99.5 %
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.01 %
- Free acid (as HCl):Max. 0.001 %
- Refractive Index:n20/D 1.505 (lit.)
- Vapor Pressure:19 mmHg ( 25 °C)
- Pakning:Glass bottle
- Pakn:6 L



